Assay Details
Gene Symbol: C22H3orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695964.3 | XP_005696021.1 | 2 - 3 | 1172 | 107 | |
XM_018066636.1 | XP_017922125.1 | 2 - 3 | 1172 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
