Assay Details
| Gene Symbol: LITAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040819.1 | XP_017896308.1 | 3 - 4 | 471 | 104 | |
| XM_018040820.1 | XP_017896309.1 | 3 - 4 | 432 | 104 | ||
| XM_018040821.1 | XP_017896310.1 | 3 - 4 | 457 | 104 | ||
More Information
Set Membership: |
|

LITAF
Genomic Map