Assay Details
| Gene Symbol: LOC102180829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043118.1 | XP_017898607.1 | 21 - 22 | 2238 | 57 | |
| XM_018043119.1 | XP_017898608.1 | 21 - 22 | 2235 | 57 | ||
| XM_018043121.1 | XP_017898610.1 | 19 - 20 | 1886 | 57 | ||
| XM_018043123.1 | XP_017898612.1 | 20 - 21 | 2053 | 57 | ||
| XM_018043124.1 | XP_017898613.1 | 20 - 21 | 1944 | 57 | ||
| XM_018043125.1 | XP_017898614.1 | 20 - 21 | 2048 | 57 | ||
| XM_018043126.1 | XP_017898615.1 | 14 - 15 | 1364 | 57 | ||
| XM_018043127.1 | XP_017898616.1 | 14 - 15 | 1303 | 57 | ||
| XM_018043128.1 | XP_017898617.1 | 13 - 14 | 1404 | 57 | ||
| XM_018043129.1 | XP_017898618.1 | 13 - 14 | 1235 | 57 | ||
| XM_018043130.1 | XP_017898619.1 | 7 - 8 | 967 | 57 | ||
| XM_018043131.1 | XP_017898620.1 | 14 - 15 | 1262 | 57 | ||
| XM_018043133.1 | XP_017898622.1 | 7 - 8 | 776 | 57 | ||
More Information
Set Membership: |
|

LOC102180829
Genomic Map