Assay Details
Gene Symbol: EPHA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041849.1 | XP_017897338.1 | 9 - 10 | 1925 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
