Assay Details
| Gene Symbol: MCAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048869.1 | XP_017904358.1 | 2 - 3 | 565 | 72 | |
| XM_018048870.1 | XP_017904359.1 | 2 - 3 | 568 | 72 | ||
More Information
Set Membership: |
|

MCAT
Genomic Map