Assay Details
| Gene Symbol: APLF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055441.1 | XP_017910930.1 | 3 - 4 | 329 | 66 | |
| XM_018055442.1 | XP_017910931.1 | 3 - 4 | 282 | 66 | ||
| XM_018055443.1 | XP_017910932.1 | 4 - 5 | 708 | 66 | ||
More Information
Set Membership: |
|

APLF
Genomic Map