Assay Details
| Gene Symbol: CDK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055822.1 | XP_017911311.1 | 6 - 7 | 1163 | 56 | |
| GenBank mRNA | JO430815.2 | - | 6 - 7 | 1034 | 56 | |
More Information
Set Membership: |
|

CDK9
Genomic Map