Assay Details
| Gene Symbol: WIPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064887.1 | XP_017920376.1 | 4 - 5 | 552 | 81 | |
| XM_018064892.1 | XP_017920381.1 | 4 - 5 | 568 | 81 | ||
More Information
Set Membership: |
|

WIPF1
Genomic Map