Assay Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061826.1 | XP_017917315.1 | 5 - 6 | 774 | 58 | |
| XM_018061827.1 | XP_017917316.1 | 4 - 5 | 687 | 58 | ||
| GenBank mRNA | JO504424.2 | - | 5 - 6 | 761 | 58 | |
More Information
Set Membership: |
|

FBXO31
Genomic Map