Assay Details
| Gene Symbol: TEAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018038905.1 | XP_017894394.1 | 11 - 12 | 1376 | 58 | |
| XM_018038906.1 | XP_017894395.1 | 11 - 12 | 1281 | 58 | ||
More Information
Set Membership: |
|

TEAD3
Genomic Map