Assay Details
| Gene Symbol: GAREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039594.1 | XP_017895083.1 | 3 - 4 | 504 | 70 | |
| XM_018039595.1 | XP_017895084.1 | 3 - 4 | 504 | 70 | ||
| GenBank mRNA | KA325078.1 | - | 3 - 4 | 468 | 70 | |
More Information
Set Membership: |
|

GAREM1
Genomic Map