Assay Details
| Gene Symbol: SDR42E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974992.2 | XP_013830446.2 | 5 - 6 | 1072 | 77 | |
| GenBank mRNA | KA333875.1 | - | 1 - 2 | 52 | 77 | |
More Information
Set Membership: |
|

SDR42E2
Genomic Map