Assay Details
| Gene Symbol: LOC102188659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044295.1 | XP_017899784.1 | 11 - 12 | 1764 | 84 | |
| GenBank mRNA | KA335395.1 | - | 1 - 2 | 180 | 84 | |
More Information
Set Membership: |
|

LOC102188659
Genomic Map