Assay Details
| Gene Symbol: LOC102188659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018044295.1 | XP_017899784.1 | 12 - 13 | 1905 | 108 | |
| GenBank mRNA | KA335395.1 | - | 2 - 3 | 321 | 108 | |
More Information
Set Membership: |
|

LOC102188659
Genomic Map