Assay Details
| Gene Symbol: CADM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674789.3 | XP_005674846.1 | 2 - 3 | 812 | 77 | |
| XM_005674791.3 | XP_005674848.1 | 3 - 4 | 839 | 77 | ||
| XM_018054195.1 | XP_017909684.1 | 2 - 3 | 218 | 77 | ||
| GenBank mRNA | JO638023.2 | - | 7 - 8 | 932 | 77 | |
| JO638024.2 | - | 7 - 8 | 932 | 77 | ||
More Information
Set Membership: |
|

CADM2
Genomic Map