Assay Details
| Gene Symbol: NDUFB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675278.3 | XP_005675335.1 | 1 - 2 | 229 | 60 | |
| GenBank mRNA | JO394522.2 | - | 5 - 6 | 1003 | 60 | |
More Information
Set Membership: |
|

NDUFB5
Genomic Map