Assay Details
| Gene Symbol: C1H3orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048605.1 | XP_017904094.1 | 4 - 5 | 511 | 98 | |
| GenBank mRNA | JT706293.1 | - | 4 - 5 | 444 | 98 | |
More Information
Set Membership: |
|

C1H3orf33
Genomic Map