Assay Details
Gene Symbol: FCRL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677243.3 | XP_005677300.2 | 7 - 8 | 1158 | 68 | |
GenBank mRNA | JO613738.2 | - | 3 - 4 | 331 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
