Assay Details
| Gene Symbol: CD209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005682411.3 | XP_005682468.1 | 2 - 3 | 169 | 75 | |
| GenBank mRNA | JO580565.1 | - | 7 - 8 | 1066 | 75 | |
More Information
Set Membership: |
|

CD209
Genomic Map