Assay Details
| Gene Symbol: GABRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683367.3 | XP_005683424.1 | 4 - 5 | 882 | 66 | |
| XM_005683368.3 | XP_005683425.1 | 4 - 5 | 882 | 66 | ||
More Information
Set Membership: |
|

GABRG2
Genomic Map