Assay Details
| Gene Symbol: TMEM252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683716.3 | XP_005683773.1 | 1 - 2 | 474 | 87 | |
| GenBank mRNA | JO427272.1 | - | 1 - 2 | 372 | 87 | |
More Information
Set Membership: |
|

TMEM252
Genomic Map