Assay Details
| Gene Symbol: MPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053459.1 | XP_017908948.1 | 4 - 5 | 433 | 76 | |
| GenBank mRNA | JO442410.2 | - | 4 - 5 | 443 | 76 | |
More Information
Set Membership: |
|

MPC1
Genomic Map