Assay Details
| Gene Symbol: MAPK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685763.3 | XP_005685820.2 | 2 - 3 | 1504 | 86 | |
| XM_018054206.1 | XP_017909695.1 | 2 - 3 | 1352 | 86 | ||
| XM_018054207.1 | XP_017909696.1 | 2 - 3 | 1340 | 86 | ||
| GenBank mRNA | JO595904.2 | - | 4 - 5 | 85 | ||
More Information
Set Membership: |
|

MAPK6
Genomic Map