Assay Details
| Gene Symbol: MRPS9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686304.3 | XP_005686361.1 | 4 - 5 | 770 | 76 | |
| GenBank mRNA | JO618368.2 | - | 4 - 5 | 487 | 76 | |
More Information
Set Membership: |
|

MRPS9
Genomic Map