Assay Details
| Gene Symbol: PTGES2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687181.3 | XP_005687238.3 | 2 - 3 | 834 | 60 | |
| GenBank mRNA | JO592648.2 | - | 5 - 6 | 1309 | 60 | |
More Information
Set Membership: |
|

PTGES2
Genomic Map