Assay Details
| Gene Symbol: TRIM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690119.3 | XP_005690176.3 | 1 - 2 | 986 | 81 | |
| XM_018059237.1 | XP_017914726.1 | 1 - 2 | 986 | 81 | ||
More Information
Set Membership: |
|

TRIM44
Genomic Map