Assay Details
| Gene Symbol: CD180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694612.2 | XP_005694669.1 | 1 - 2 | 217 | 96 | |
| GenBank mRNA | JO577905.1 | - | 1 - 2 | 301 | 96 | |
More Information
Set Membership: |
|

CD180
Genomic Map