Assay Details
| Gene Symbol: FYB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694771.3 | XP_005694828.2 | 10 - 11 | 1989 | 94 | |
| XM_005694772.3 | XP_005694829.2 | 10 - 11 | 1988 | 94 | ||
| GenBank mRNA | JO579042.1 | - | 9 - 10 | 1781 | 94 | |
More Information
Set Membership: |
|

FYB1
Genomic Map