Assay Details
| Gene Symbol: SLC39A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696976.2 | XP_005697033.2 | 9 - 10 | 2825 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC39A6| Gene Symbol: SLC39A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005696976.2 | XP_005697033.2 | 9 - 10 | 2825 | 60 | |
Set Membership: |
|