Assay Details
| Gene Symbol: POLI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039699.1 | XP_017895188.1 | 4 - 5 | 1023 | 68 | |
| XM_018039700.1 | XP_017895189.1 | 4 - 5 | 955 | 68 | ||
| XM_018039701.1 | XP_017895190.1 | 3 - 4 | 455 | 68 | ||
More Information
Set Membership: |
|

POLI
Genomic Map