Assay Details
| Gene Symbol: CSGALNACT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042390.1 | XP_017897879.1 | 3 - 4 | 1303 | 89 | |
| XM_018042391.1 | XP_017897880.1 | 3 - 4 | 1303 | 89 | ||
| XM_018042392.1 | XP_017897881.1 | 3 - 4 | 1303 | 89 | ||
| XM_018042393.1 | XP_017897882.1 | 3 - 4 | 1303 | 89 | ||
| GenBank mRNA | JT753722.1 | - | 5 - 6 | 1132 | 89 | |
More Information
Set Membership: |
|

CSGALNACT2
Genomic Map