Assay Details
| Gene Symbol: SRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005699128.3 | XP_005699185.1 | 1 - 2 | 167 | 70 | |
| XM_005699129.3 | XP_005699186.1 | 1 - 2 | 161 | 70 | ||
| GenBank mRNA | JO581015.1 | - | 2 - 3 | 512 | 70 | |
More Information
Set Membership: |
|

SRGN
Genomic Map