Assay Details
| Gene Symbol: ITPRID1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018047120.1 | XP_017902609.1 | 12 - 13 | 2790 | 64 | |
| XM_018047121.1 | XP_017902610.1 | 12 - 13 | 2790 | 64 | ||
| GenBank mRNA | JT640580.1 | - | 4 - 5 | 676 | 64 | |
More Information
Set Membership: |
|

ITPRID1
Genomic Map