Assay Details
| Gene Symbol: NMNAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049838.1 | XP_017905327.1 | 2 - 3 | 550 | 100 | |
| GenBank mRNA | JO454810.2 | - | 2 - 3 | 255 | 100 | |
More Information
Set Membership: |
|

NMNAT3
Genomic Map