Assay Details
| Gene Symbol: NR6A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055752.1 | XP_017911241.1 | 6 - 7 | 978 | 61 | |
| XM_018055753.1 | XP_017911242.1 | 6 - 7 | 822 | 61 | ||
| GenBank mRNA | JT680650.1 | - | 4 - 5 | 475 | 61 | |
More Information
Set Membership: |
|

NR6A1
Genomic Map