Assay Details
| Gene Symbol: ARHGAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059139.1 | XP_017914628.1 | 6 - 7 | 917 | 59 | |
| XM_018059140.1 | XP_017914629.1 | 5 - 6 | 878 | 59 | ||
| XM_018059141.1 | XP_017914630.1 | 6 - 7 | 641 | 59 | ||
| XM_018059142.1 | XP_017914631.1 | 5 - 6 | 611 | 59 | ||
| XM_018059143.1 | XP_017914632.1 | 5 - 6 | 646 | 59 | ||
| GenBank mRNA | JO613226.2 | - | 5 - 6 | 614 | 59 | |
| KA318736.1 | - | 8 - 9 | 2461 | 59 | ||
More Information
Set Membership: |
|

ARHGAP1
Genomic Map