Assay Details
| Gene Symbol: TP53BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060196.1 | XP_017915685.1 | 13 - 14 | 3024 | 61 | |
| XM_018060199.1 | XP_017915688.1 | 13 - 14 | 3012 | 61 | ||
| XM_018060200.1 | XP_017915689.1 | 12 - 13 | 2721 | 61 | ||
| XM_018060201.1 | XP_017915690.1 | 12 - 13 | 2913 | 61 | ||
More Information
Set Membership: |
|

TP53BP2
Genomic Map