Assay Details
| Gene Symbol: HHAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060770.1 | XP_017916259.1 | 4 - 5 | 468 | 56 | |
| XM_018060771.1 | XP_017916260.1 | 4 - 5 | 468 | 56 | ||
| GenBank mRNA | JO647544.2 | - | 1 - 2 | 130 | 56 | |
More Information
Set Membership: |
|

HHAT
Genomic Map