Assay Details
| Gene Symbol: RNF215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060990.1 | XP_017916479.1 | 3 - 4 | 557 | 65 | |
| XM_018060991.1 | XP_017916480.1 | 3 - 4 | 443 | 65 | ||
| GenBank mRNA | JO357178.2 | - | 2 - 3 | 162 | 65 | |
More Information
Set Membership: |
|

RNF215
Genomic Map