Assay Details
| Gene Symbol: TOGARAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973177.2 | XP_013828631.2 | 11 - 12 | 4014 | 107 | |
| XM_013973178.2 | XP_013828632.2 | 10 - 11 | 3861 | 107 | ||
| XM_018066410.1 | XP_017921899.1 | 11 - 12 | 4014 | 107 | ||
More Information
Set Membership: |
|

TOGARAM1
Genomic Map