Assay Details
| Gene Symbol: TOGARAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973177.2 | XP_013828631.2 | 12 - 13 | 4162 | 91 | |
| XM_013973178.2 | XP_013828632.2 | 11 - 12 | 4009 | 91 | ||
| XM_018066410.1 | XP_017921899.1 | 12 - 13 | 4162 | 91 | ||
More Information
Set Membership: |
|

TOGARAM1
Genomic Map