Assay Details
| Gene Symbol: TOGARAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973177.2 | XP_013828631.2 | 18 - 19 | 5093 | 107 | |
| XM_013973178.2 | XP_013828632.2 | 17 - 18 | 4940 | 107 | ||
More Information
Set Membership: |
|

TOGARAM1
Genomic Map