Assay Details
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041611.1 | XP_017897100.1 | 3 - 4 | 397 | 64 | |
GenBank mRNA | JO354766.2 | - | 2 - 3 | 397 | 64 | |
JO613399.2 | - | 2 - 3 | 336 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
