Assay Details
| Gene Symbol: HELT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041818.1 | XP_017897307.1 | 2 - 3 | 216 | 71 | |
| XM_018041819.1 | XP_017897308.1 | 2 - 3 | 216 | 71 | ||
More Information
Set Membership: |
|

HELT
Genomic Map