Assay Details
Gene Symbol: LOC102175808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675472.2 | XP_005675529.1 | 5 - 6 | 678 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
