Assay Details
| Gene Symbol: CCL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676644.3 | XP_005676701.2 | 2 - 3 | 275 | 71 | |
| XM_005676645.3 | XP_005676702.2 | 2 - 3 | 272 | 71 | ||
More Information
Set Membership: |
|

CCL20
Genomic Map