Assay Details
| Gene Symbol: DBF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679056.3 | XP_005679113.2 | 6 - 7 | 858 | 75 | |
| XM_005679057.3 | XP_005679114.2 | 6 - 7 | 859 | 75 | ||
| XM_018047308.1 | XP_017902797.1 | 5 - 6 | 681 | 75 | ||
| GenBank mRNA | JT687969.1 | - | 9 - 10 | 1237 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018047308.1 transcripts.
Set Membership: |
|

DBF4
Genomic Map