Assay Details
Gene Symbol: SYCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682278.3 | XP_005682335.1 | 2 - 3 | 222 | 68 | |
XM_005682280.3 | XP_005682337.1 | 2 - 3 | 234 | 68 | ||
GenBank mRNA | JO588909.2 | - | 3 - 4 | 747 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
