Assay Details
| Gene Symbol: SLC23A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683010.3 | XP_005683067.1 | 14 - 15 | 2266 | 63 | |
| XM_013965597.2 | XP_013821051.1 | 14 - 15 | 2267 | 63 | ||
| XM_013965598.2 | XP_013821052.1 | 14 - 15 | 2267 | 63 | ||
| XM_013965599.2 | XP_013821053.1 | 15 - 16 | 1786 | 63 | ||
| XM_018050309.1 | XP_017905798.1 | 14 - 15 | 1719 | 63 | ||
| XM_018050310.1 | XP_017905799.1 | 15 - 16 | 1822 | 63 | ||
| XM_018050311.1 | XP_017905800.1 | 12 - 13 | 1963 | 63 | ||
More Information
Set Membership: |
|

SLC23A1
Genomic Map