Assay Details
| Gene Symbol: F2RL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685060.3 | XP_005685117.2 | 1 - 2 | 373 | 59 | |
| GenBank mRNA | JO429978.2 | - | 1 - 2 | 1795 | 59 | |
| KA314847.1 | - | 284 | 59 | |||
More Information
Set Membership: |
|

F2RL2
Genomic Map